siRNA Details
| Gene Symbol: Nkd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163660.1 | NP_001157132.1 | 8 | 978 | ||
| NM_027280.3 | NP_081556.3 | 10 | 1188 | |||
| GenBank mRNA | AF343352.1 | - | 10 | 1025 | ||
| AF358134.1 | - | 10 | 1104 | |||
| AK076164.1 | - | 10 | 1179 | |||
| AK082367.1 | - | 10 | 1177 | |||
| BC034838.1 | - | 10 | 1162 | 1429943 | ||

Nkd1
Genomic Map