siRNA Details
| Gene Symbol: Hemgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053149.2 | NP_444379.1 | 3 | 1296 | ||
| GenBank mRNA | AF269248.1 | - | 3 | 1296 | ||
| AK076587.1 | - | 4 | 1281 | |||
| AK077692.1 | - | 3 | 1139 | |||
| AK156536.1 | - | 3 | 1224 | |||
| AK171890.1 | - | 3 | 1215 | |||
| AK171906.1 | - | 3 | 1350 | |||
| AK172421.1 | - | 3 | 1216 | |||
| AY699986.1 | - | 3 | 1138 | |||
| BC050798.1 | - | 4 | 1275 | 6743819 | ||

Hemgn
Genomic Map