siRNA Details
| Gene Symbol: Klra20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053150.2 | NP_444380.1 | 5 | 651 | ||
| GenBank mRNA | AF288379.1 | - | 5 | 651 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klra20| Gene Symbol: Klra20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053150.2 | NP_444380.1 | 5 | 651 | ||
| GenBank mRNA | AF288379.1 | - | 5 | 651 | ||