siRNA Details
| Gene Symbol: Klra22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053152.2 | NP_444382.1 | 6 | 729 | ||
| GenBank mRNA | AF288381.1 | - | 6 | 729 | ||
| Gene Symbol: Klra1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF419250.1 | - | 7 | 1068 | ||
| AF444273.1 | - | 7 | 1068 | |||

Klra22
Genomic Map