siRNA Details
| Gene Symbol: Tm2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053157.2 | NP_444387.1 | 6 | 576 | ||
| GenBank mRNA | AF353993.1 | - | 6 | 550 | ||
| AK009511.1 | - | 6 | 540 | |||
| BC089492.1 | - | 6 | 576 | 6703812 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tm2d1| Gene Symbol: Tm2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053157.2 | NP_444387.1 | 6 | 576 | ||
| GenBank mRNA | AF353993.1 | - | 6 | 550 | ||
| AK009511.1 | - | 6 | 540 | |||
| BC089492.1 | - | 6 | 576 | 6703812 | ||