siRNA Details
| Gene Symbol: Bcl2l13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153516.2 | NP_705736.1 | 7 | 838 | ||
| GenBank mRNA | AK031018.1 | - | 7 | 787 | ||
| AK047223.1 | - | 7 | 840 | |||
| AK088417.1 | - | 7 | 853 | |||
| AK132774.1 | - | 7 | 847 | |||
| AK143457.1 | - | 7 | 853 | |||
| AK148007.1 | - | 7 | 857 | |||
| AK165068.1 | - | 7 | 831 | |||
| AK168833.1 | - | 7 | 828 | |||
| BC027307.1 | - | 7 | 728 | 4014844 | ||
| BC029016.1 | - | 7 | 838 | 5358785 | ||

Bcl2l13
Genomic Map