siRNA Details
| Gene Symbol: Mrpl43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053164.3 | NP_444394.1 | 2 | 235 | ||
| GenBank mRNA | AB049657.1 | - | 2 | 196 | ||
| AK015357.1 | - | 2 | 216 | |||
| AK076204.1 | - | 2 | 194 | |||
| BC027676.1 | - | 2 | 175 | |||
| BC048733.1 | - | 2 | 226 | 6512868 | ||
| BC065812.1 | - | 2 | 182 | |||

Mrpl43
Genomic Map