siRNA Details
| Gene Symbol: Trim6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013616.2 | NP_001013637.1 | 2 | 268 | ||
| GenBank mRNA | AK033396.1 | - | 2 | 248 | ||
| AK040000.1 | - | 2 | 271 | |||
| AK049238.1 | - | 2 | 248 | |||
| BB634731.1 | - | 2 | 268 | |||
| BC138965.1 | - | 2 | 199 | 8861987 | ||

Trim6
Genomic Map