siRNA Details
| Gene Symbol: Trim16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053169.2 | NP_444399.2 | 6 | 1573 | ||
| GenBank mRNA | AF220134.1 | - | 2 | 291 | ||
| AF449496.1 | - | 6 | 1324 | |||
| AK053139.1 | - | 6 | 1387 | |||
| AK078902.1 | - | 4 | 909 | |||
| BC052821.1 | - | 7 | 1275 | 30057830 | ||

Trim16
Genomic Map