siRNA Details
| Gene Symbol: Trim33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001079830.2 | NP_001073299.1 | 17 | 3549 | ||
| NM_053170.3 | NP_444400.2 | 17 | 3549 | |||
| GenBank mRNA | AF220138.1 | - | 2 | 126 | ||
| AK129293.1 | - | 17 | 2808 | |||
| AY458590.1 | - | 17 | 3555 | |||
| BC043471.1 | - | 4 | 544 | |||

Trim33
Genomic Map