siRNA Details
| Gene Symbol: NPTX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002523.2 | NP_002514.1 | 5 | 2175 | ||
| GenBank mRNA | BC009924.2 | - | 5 | 2167 | 3508049 | |
| BC034781.1 | - | 5 | 2163 | 5169124 | ||
| BC035339.1 | - | 5 | 2168 | 5198692 | ||
| BC048275.1 | - | 5 | 2133 | 5165672 | ||
| U26662.1 | - | 5 | 1889 | |||

NPTX2
Genomic Map