siRNA Details
| Gene Symbol: Csmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053171.2 | NP_444401.2 | 28 | 4845 | ||
| GenBank mRNA | AK081081.1 | - | 16 | 2846 | ||
| AY017475.1 | - | 28 | 4742 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csmd1| Gene Symbol: Csmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053171.2 | NP_444401.2 | 28 | 4845 | ||
| GenBank mRNA | AK081081.1 | - | 16 | 2846 | ||
| AY017475.1 | - | 28 | 4742 | |||