siRNA Details
| Gene Symbol: Csmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053171.2 | NP_444401.2 | 35 | 5908 | ||
| GenBank mRNA | AY017475.1 | - | 35 | 5805 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csmd1| Gene Symbol: Csmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053171.2 | NP_444401.2 | 35 | 5908 | ||
| GenBank mRNA | AY017475.1 | - | 35 | 5805 | ||