siRNA Details
| Gene Symbol: NPTX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002523.2 | NP_002514.1 | 3, 4 | 1058 | ||
| GenBank mRNA | AK311250.1 | - | 2, 3 | 749 | ||
| BC009924.2 | - | 3, 4 | 1050 | 3508049 | ||
| BC034781.1 | - | 3, 4 | 1046 | 5169124 | ||
| BC035339.1 | - | 3, 4 | 1051 | 5198692 | ||
| BC048275.1 | - | 3, 4 | 1016 | 5165672 | ||
| U26662.1 | - | 3, 4 | 774 | |||

NPTX2
Genomic Map