siRNA Details
| Gene Symbol: Zfp423 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033327.2 | NP_201584.2 | 4 | 1743 | ||
| GenBank mRNA | AF188609.1 | - | 2 | 1112 | ||
| AK122365.1 | - | 4 | 1709 | |||
| AK133497.1 | - | 2 | 1171 | |||
| AY147407.1 | - | 3 | 1348 | |||
| AY256893.1 | - | 4 | 1642 | |||
| BC059234.1 | - | 2 | 1554 | |||
| BC079586.1 | - | 4 | 1429 | |||
| BC139028.1 | - | 4 | 1489 | 8862050 | ||
| BC139030.1 | - | 4 | 1489 | 8862052 | ||

Zfp423
Genomic Map