siRNA Details
| Gene Symbol: Ophn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052976.3 | NP_443208.1 | 5 | 954 | ||
| GenBank mRNA | AK028822.1 | - | 5 | 438 | ||
| AK031419.1 | - | 5 | 947 | |||
| AK036038.1 | - | 5 | 955 | |||
| AK039304.1 | - | 4 | 1122 | |||
| AK046214.1 | - | 5 | 410 | |||
| AK048153.1 | - | 5 | 955 | |||
| AK082551.1 | - | 5 | 594 | |||
| AK087469.1 | - | 5 | 418 | |||
| BC004845.1 | - | 6 | 552 | 2650009 | ||

Ophn1
Genomic Map