siRNA Details
| Gene Symbol: Pag1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001195031.1 | NP_001181960.1 | 7 | 1459 | ||
| NM_053182.5 | NP_444412.2 | 6 | 1400 | |||
| GenBank mRNA | AF250192.1 | - | 4 | 781 | ||
| AK040230.1 | - | 7 | 1409 | |||
| AK076515.1 | - | 3 | 898 | |||
| AK089329.1 | - | 6 | 1352 | |||
| AK156000.1 | - | 7 | 1411 | |||
| AK156986.1 | - | 7 | 1383 | |||
| AK172106.1 | - | 7 | 1382 | |||
| BC145328.1 | - | 7 | 1283 | 9053884 | ||
| BC145761.1 | - | 7 | 1283 | 40131064 | ||
| BC145763.1 | - | 7 | 1283 | 40131066 | ||

Pag1
Genomic Map