siRNA Details
| Gene Symbol: Ugt2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053184.2 | NP_444414.2 | 2 | 777 | ||
| GenBank mRNA | AF184901.1 | - | 1 | 693 | ||
| AK140757.1 | - | 2 | 778 | |||
| BC048926.1 | - | 1 | 765 | 6314878 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ugt2a1| Gene Symbol: Ugt2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053184.2 | NP_444414.2 | 2 | 777 | ||
| GenBank mRNA | AF184901.1 | - | 1 | 693 | ||
| AK140757.1 | - | 2 | 778 | |||
| BC048926.1 | - | 1 | 765 | 6314878 | ||