siRNA Details
| Gene Symbol: Cnnm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039551.1 | NP_001034640.1 | 1 | 1237 | ||
| NM_053186.2 | NP_444416.2 | 1 | 1237 | |||
| GenBank mRNA | AF216964.1 | - | 1 | 306 | ||
| BC052714.1 | - | 1 | 307 | |||
| BC080275.1 | - | 1 | 664 | |||
| BC108417.1 | - | 1 | 491 | |||
| BC132285.1 | - | 1 | 289 | |||
| BC144720.1 | - | 1 | 288 | |||
| GQ225746.2 | - | 1 | 223 | |||

Cnnm3
Genomic Map