siRNA Details
| Gene Symbol: Srd5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053188.2 | NP_444418.1 | 1, 2 | 362 | ||
| GenBank mRNA | AB049456.1 | - | 1, 2 | 362 | ||
| AK138946.1 | - | 1, 2 | 303 | |||
| BC125510.1 | - | 1, 2 | 346 | 40130025 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Srd5a2| Gene Symbol: Srd5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053188.2 | NP_444418.1 | 1, 2 | 362 | ||
| GenBank mRNA | AB049456.1 | - | 1, 2 | 362 | ||
| AK138946.1 | - | 1, 2 | 303 | |||
| BC125510.1 | - | 1, 2 | 346 | 40130025 | ||