siRNA Details
| Gene Symbol: Pi15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053191.2 | NP_444421.2 | 3 | 522 | ||
| GenBank mRNA | AF329196.1 | - | 3 | 442 | ||
| AK040961.1 | - | 3 | 521 | |||
| AK165584.1 | - | 2 | 198 | |||
| BC116705.1 | - | 3 | 418 | 40102877 | ||

Pi15
Genomic Map