siRNA Details
| Gene Symbol: Fkbp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277891.1 | NP_001264820.1 | 2, 3 | 225 | ||
| NM_001277892.1 | NP_001264821.1 | 2, 3 | 172 | |||
| NM_001277893.1 | NP_001264822.1 | 2, 3 | 225 | |||
| NM_033571.3 | NP_291049.1 | 2, 3 | 225 | |||
| GenBank mRNA | AF367709.1 | - | 2, 3 | 206 | ||
| AF367710.1 | - | 2, 3 | 172 | |||
| AF367711.1 | - | 2, 3 | 172 | |||
| AK090036.1 | - | 2, 3 | 216 | |||
| AY029192.1 | - | 2, 3 | 172 | |||
| BC139092.1 | - | 2, 3 | 225 | 8862114 | ||
| BC139097.1 | - | 2, 3 | 225 | 8862119 | ||

Fkbp6
Genomic Map