siRNA Details
| Gene Symbol: Prcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033573.2 | NP_291051.1 | 6 | 1586 | ||
| GenBank mRNA | AF273750.1 | - | 6 | 1607 | ||
| AK078394.1 | - | 6 | 1606 | |||
| BC052072.1 | - | 6 | 1586 | 5717482 | ||
| BC082784.1 | - | 6 | 1604 | 4191155 | ||

Prcc
Genomic Map