siRNA Details
| Gene Symbol: Cadm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053199.3 | NP_444429.1 | 4 | 601 | ||
| GenBank mRNA | AF195662.1 | - | 4 | 545 | ||
| AK036352.1 | - | 5 | 657 | |||
| AK038917.1 | - | 4 | 558 | |||
| AK053077.1 | - | 4 | 555 | |||
| AY059393.1 | - | 4 | 400 | |||
| BC029659.1 | - | 1 | 20 | |||
| BC079837.1 | - | 5 | 641 | |||
| BC115417.2 | - | 4 | 506 | 40093410 | ||
| BC115418.2 | - | 4 | 506 | 40093417 | ||

Cadm3
Genomic Map