siRNA Details
| Gene Symbol: Cadm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053199.3 | NP_444429.1 | 2 | 304 | ||
| GenBank mRNA | AF195662.1 | - | 2 | 248 | ||
| AK036352.1 | - | 3 | 360 | |||
| AK038917.1 | - | 2 | 261 | |||
| AK053077.1 | - | 2 | 258 | |||
| AY059393.1 | - | 2 | 103 | |||
| BC079837.1 | - | 3 | 344 | |||
| BC115417.2 | - | 2 | 209 | 40093410 | ||
| BC115418.2 | - | 2 | 209 | 40093417 | ||
| BY289385.1 | - | 2 | 306 | |||

Cadm3
Genomic Map