siRNA Details
| Gene Symbol: Tprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175286.4 | NP_780495.2 | 1, 2 | 1963 | ||
| GenBank mRNA | AK082888.1 | - | 1, 2 | 1944 | ||
| AK171799.1 | - | 1, 2 | 1977 | |||
| BC024573.1 | - | 1, 2 | 268 | |||
| BC055478.1 | - | 1, 2 | 408 | |||
| BC089508.1 | - | 1, 2 | 1473 | 6589951 | ||

Tprn
Genomic Map