siRNA Details
| Gene Symbol: Nmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133787.2 | NP_598548.1 | 13, 14 | 1285 | ||
| GenBank mRNA | AK034772.1 | - | 485 | |||
| AK088667.1 | - | 13, 14 | 1298 | |||
| AK145248.1 | - | 13, 14 | 1307 | |||
| AK166283.1 | - | 13, 14 | 1253 | |||
| BC003842.1 | - | 13, 14 | 1285 | 3495021 | ||

Nmd3
Genomic Map