siRNA Details
| Gene Symbol: Cog8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139229.4 | NP_631975.3 | 3 | 1238 | ||
| GenBank mRNA | AB041610.2 | - | 3 | 1161 | ||
| AK049957.1 | - | 3 | 1186 | |||
| AK083734.1 | - | 3 | 1180 | |||
| AK145515.1 | - | 3 | 1191 | |||
| AK145727.1 | - | 3 | 1182 | |||
| AK153617.1 | - | 3 | 1182 | |||
| AK156186.1 | - | 3 | 1181 | |||
| AK159569.1 | - | 3 | 1179 | |||
| BC094654.1 | - | 1174 | 6853777 | |||

Cog8
Genomic Map