siRNA Details
| Gene Symbol: 4833439L19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252645.1 | NP_001239574.1 | 5, 6 | 736 | ||
| NM_001252646.1 | NP_001239575.1 | 4, 5 | 629 | |||
| NM_001252647.1 | NP_001239576.1 | 3, 4 | 450 | |||
| NM_001252648.1 | NP_001239577.1 | 2, 3 | 323 | |||
| NM_133797.4 | NP_598558.1 | 6, 7 | 824 | |||
| GenBank mRNA | AK004845.1 | - | 6, 7 | 783 | ||
| AK014792.1 | - | 2, 3 | 257 | |||
| AK028697.1 | - | 6, 7 | 757 | |||
| AK145025.1 | - | 6, 7 | 790 | |||
| AK159945.1 | - | 6, 7 | 787 | |||
| AK167616.1 | - | 6, 7 | 768 | |||
| AK169399.1 | - | 6, 7 | 790 | |||
| AK173126.1 | - | 6, 7 | 737 | |||
| BC033445.1 | - | 6, 7 | 803 | 4222839 | ||
| BC039775.1 | - | 6, 7 | 762 | 4239147 | ||
| BC043040.1 | - | 6, 7 | 780 | 5701249 | ||
| BC054417.1 | - | 6, 7 | 769 | 3969623 | ||
| BY244215.1 | - | 3, 4 | 413 | |||
| BY265900.1 | - | 2, 3 | 324 | |||
| CA461006.1 | - | 4, 5 | 631 | |||

4833439L19Rik
Genomic Map