siRNA Details
| Gene Symbol: Nlrp4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175290.4 | NP_780499.3 | 3 | 1687 | ||
| GenBank mRNA | AK049352.1 | - | 1 | 90 | ||
| AY596201.1 | - | 4 | 1797 | |||
| BC098479.1 | - | 1 | 109 | |||
| DV656119.1 | - | 1 | 715 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nlrp4f| Gene Symbol: Nlrp4f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175290.4 | NP_780499.3 | 3 | 1687 | ||
| GenBank mRNA | AK049352.1 | - | 1 | 90 | ||
| AY596201.1 | - | 4 | 1797 | |||
| BC098479.1 | - | 1 | 109 | |||
| DV656119.1 | - | 1 | 715 | |||