siRNA Details
| Gene Symbol: Gtf2f1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133801.2 | NP_598562.1 | 4 | 379 | ||
| GenBank mRNA | AK013002.1 | - | 4 | 393 | ||
| AK078048.1 | - | 4 | 392 | |||
| AK168236.1 | - | 4 | 392 | |||
| AK168243.1 | - | 4 | 392 | |||
| BC003336.1 | - | 4 | 377 | |||
| BC023081.1 | - | 4 | 379 | 5355475 | ||
| BC031123.1 | - | 4 | 509 | 5026448 | ||

Gtf2f1
Genomic Map