siRNA Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153555.2 | NP_705783.1 | 10, 11 | 1531 | ||
| GenBank mRNA | AK035701.1 | - | 10, 11 | 1430 | ||
| AK098114.1 | - | 10, 11 | 1550 | |||
| AK146825.1 | - | 10, 11 | 1458 | |||
| AK162622.1 | - | 8, 9 | 2479 | |||
| AK167059.1 | - | 10, 11 | 1407 | |||
| BC023704.1 | - | 10, 11 | 1414 | 5345701 | ||
| BC023804.1 | - | 11, 12 | 1585 | 5352941 | ||
| BC078641.1 | - | 10, 11 | 2443 | |||

Dcaf8
Genomic Map