siRNA Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153555.2 | NP_705783.1 | 6, 7 | 1195 | ||
| GenBank mRNA | AK035701.1 | - | 6, 7 | 1094 | ||
| AK098114.1 | - | 6, 7 | 1214 | |||
| AK146825.1 | - | 6, 7 | 1122 | |||
| AK162622.1 | - | 6, 7 | 1217 | |||
| AK164799.1 | - | 6, 7 | 1122 | |||
| AK167059.1 | - | 6, 7 | 1071 | |||
| BC023704.1 | - | 7, 8 | 1136 | 5345701 | ||
| BC023804.1 | - | 7, 8 | 1249 | 5352941 | ||
| BC078641.1 | - | 6, 7 | 2107 | |||

Dcaf8
Genomic Map