siRNA Details
| Gene Symbol: Dcaf8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153555.2 | NP_705783.1 | 7, 8 | 1292 | ||
| GenBank mRNA | AK035701.1 | - | 7, 8 | 1191 | ||
| AK098114.1 | - | 7, 8 | 1311 | |||
| AK146825.1 | - | 7, 8 | 1219 | |||
| AK162622.1 | - | 7, 8 | 1314 | |||
| AK164799.1 | - | 7, 8 | 1219 | |||
| AK167059.1 | - | 7, 8 | 1168 | |||
| AK194034.1 | - | 1, 2 | 95 | |||
| BC023704.1 | - | 8, 9 | 1233 | 5345701 | ||
| BC023804.1 | - | 8, 9 | 1346 | 5352941 | ||
| BC078641.1 | - | 7, 8 | 2204 | |||

Dcaf8
Genomic Map