siRNA Details
| Gene Symbol: Kmo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133809.1 | NP_598570.1 | 3 | 203 | ||
| GenBank mRNA | AK129011.1 | - | 216 | |||
| AK133074.1 | - | 311 | ||||
| AK158251.1 | - | 3 | 398 | |||
| AK171812.1 | - | 3 | 327 | |||
| BC014683.1 | - | 3 | 203 | 4210278 | ||

Kmo
Genomic Map