siRNA Details
| Gene Symbol: NRGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126181.1 | NP_001119653.1 | 2 | 193 | ||
| NM_006176.2 | NP_006167.1 | 2 | 193 | |||
| GenBank mRNA | BC002835.2 | - | 2 | 81 | 3660038 | |
| BM923818.1 | - | 2 | 136 | |||
| BT007325.1 | - | 1 | 41 | |||
| U89165.1 | - | 2 | 181 | |||
| Y09689.1 | - | 2 | 170 | |||

NRGN
Genomic Map