siRNA Details
| Gene Symbol: Lbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133815.2 | NP_598576.2 | 4 | 593 | ||
| GenBank mRNA | AK028426.1 | - | 4 | 570 | ||
| AK030606.1 | - | 4 | 547 | |||
| AK033459.1 | - | 4 | 545 | |||
| AK088179.1 | - | 4 | 546 | |||
| AK151798.1 | - | 4 | 560 | |||
| AK161762.1 | - | 4 | 571 | |||
| AK166850.1 | - | 4 | 533 | |||
| AK167157.1 | - | 4 | 538 | |||
| AK199017.1 | - | 1 | 80 | |||
| BC021516.1 | - | 4 | 534 | 5352337 | ||
| BC025583.1 | - | 3 | 343 | |||
| BC042522.1 | - | 4 | 553 | 4216442 | ||

Lbr
Genomic Map