siRNA Details
| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290699.1 | NP_001277628.1 | 11 | 3046 | ||
| NM_133817.3 | NP_598578.1 | 13 | 3284 | |||
| GenBank mRNA | AK030088.1 | - | 13 | 3117 | ||
| BC022767.1 | - | 1 | 90 | |||
| BC024435.1 | - | 4 | 959 | |||
| BC026587.1 | - | 4 | 1462 | |||
| BC043685.1 | - | 4 | 1534 | |||
| BC062154.1 | - | 11 | 2836 | 6331597 | ||

Zfp451
Genomic Map