siRNA Details
| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133817.3 | NP_598578.1 | 4 | 445 | ||
| GenBank mRNA | AK030088.1 | - | 4 | 278 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp451| Gene Symbol: Zfp451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133817.3 | NP_598578.1 | 4 | 445 | ||
| GenBank mRNA | AK030088.1 | - | 4 | 278 | ||