siRNA Details
| Gene Symbol: Celrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045543.1 | - | 54 | |||
| AK138346.1 | - | 1 | 122 | |||
| BE654424.1 | - | 1 | 32 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Celrr| Gene Symbol: Celrr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK045543.1 | - | 54 | |||
| AK138346.1 | - | 1 | 122 | |||
| BE654424.1 | - | 1 | 32 | |||