siRNA Details
| Gene Symbol: Pid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001003948.2 | NP_001003948.2 | 3 | 877 | ||
| GenBank mRNA | AK035542.1 | - | 3 | 752 | ||
| AK083256.1 | - | 3 | 798 | |||
| AK150971.1 | - | 3 | 815 | |||
| AK151267.1 | - | 3 | 847 | |||
| BC054112.1 | - | 2 | 510 | |||
| BC080290.1 | - | 3 | 823 | 6816865 | ||

Pid1
Genomic Map