siRNA Details
| Gene Symbol: ACTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104.3 | NP_001095.2 | 4, 5 | 558 | ||
| NM_001258371.2 | NP_001245300.2 | 4, 5 | 706 | |||
| NR_047663.2 | 4, 5 | 558 | ||||
| GenBank mRNA | AK125851.1 | - | 4, 5 | 1092 | ||
| AK303044.1 | - | 4, 5 | 706 | |||
| AK313556.1 | - | 4, 5 | 496 | |||
| BC099647.3 | - | 4, 5 | 479 | 40008220 | ||
| BC099649.3 | - | 4, 5 | 479 | 40008223 | ||
| M86407.1 | - | 4, 5 | 479 | |||

ACTN3
Genomic Map