siRNA Details
| Gene Symbol: Atp1a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178405.3 | NP_848492.1 | 6, 7 | 811 | ||
| GenBank mRNA | AK052863.1 | - | 6, 7 | 748 | ||
| AK129212.1 | - | 6, 7 | 724 | |||
| AK137602.1 | - | 6, 7 | 747 | |||
| AK147331.1 | - | 6, 7 | 743 | |||
| AK148775.1 | - | 6, 7 | 743 | |||
| AK161840.1 | - | 6, 7 | 743 | |||
| AK164509.1 | - | 6, 7 | 747 | |||
| AK168703.1 | - | 6, 7 | 743 | |||
| BC036127.1 | - | 6, 7 | 735 | 4955003 | ||
| BC041774.1 | - | 3, 4 | 461 | |||

Atp1a2
Genomic Map