siRNA Details
| Gene Symbol: Obsl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178884.5 | NP_849215.3 | 7 | 2788 | ||
| GenBank mRNA | AK029863.1 | - | 7 | 2577 | ||
| AK048144.1 | - | 7 | 1980 | |||
| AK122340.1 | - | 1 | 22 | |||
| AK162473.1 | - | 7 | 2698 | |||
| BC051643.1 | - | 5 | 1340 | |||
| BC056205.1 | - | 4 | 962 | |||
| CA750706.1 | - | 3 | 707 | |||

Obsl1
Genomic Map