siRNA Details
| Gene Symbol: Ubac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133835.2 | NP_598596.2 | 2 | 460 | ||
| GenBank mRNA | AK157857.1 | - | 2 | 294 | ||
| BC021811.1 | - | 2 | 411 | 3662474 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ubac1| Gene Symbol: Ubac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133835.2 | NP_598596.2 | 2 | 460 | ||
| GenBank mRNA | AK157857.1 | - | 2 | 294 | ||
| BC021811.1 | - | 2 | 411 | 3662474 | ||