siRNA Details
| Gene Symbol: Cdc123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133837.4 | NP_598598.1 | 9 | 714 | ||
| GenBank mRNA | AK090122.1 | - | 9 | 715 | ||
| AK141463.1 | - | 9 | 702 | |||
| AK159208.1 | - | 9 | 735 | |||
| BC019753.1 | - | 9 | 689 | 5008314 | ||
| BC022972.1 | - | 9 | 703 | 5368775 | ||
| BC023821.1 | - | 9 | 706 | 5354711 | ||
| BC024787.1 | - | 9 | 701 | 5359215 | ||

Cdc123
Genomic Map