siRNA Details
| Gene Symbol: AI182371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243102.1 | NP_001230031.1 | 5, 6 | 649 | ||
| NM_001243103.1 | NP_001230032.1 | 5, 6 | 646 | |||
| NM_178885.4 | NP_849216.2 | 5, 6 | 646 | |||
| GenBank mRNA | AK050126.1 | - | 5, 6 | 650 | ||
| AK050413.1 | - | 5, 6 | 621 | |||
| BC019541.1 | - | 5, 6 | 604 | |||
| BC037995.1 | - | 5, 6 | 626 | |||
| BC094504.1 | - | 5, 6 | 634 | 5100182 | ||
| BC115979.1 | - | 5, 6 | 621 | 40104352 | ||
| BC115986.1 | - | 5, 6 | 621 | 40104349 | ||

AI182371
Genomic Map