siRNA Details
| Gene Symbol: Myl9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172118.1 | NP_742116.1 | 2 | 72 | ||
| GenBank mRNA | AK007972.1 | - | 2 | 82 | ||
| AK020258.1 | - | 2 | 73 | |||
| AK159880.1 | - | 2 | 117 | |||
| BC049974.1 | - | 2 | 107 | 4912899 | ||
| BC055439.1 | - | 2 | 96 | 2644903 | ||

Myl9
Genomic Map