siRNA Details
| Gene Symbol: Fibcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178887.4 | NP_849218.2 | 6 | 1212 | ||
| GenBank mRNA | AK046338.1 | - | 6 | 1174 | ||
| AK084379.1 | - | 7 | 1215 | |||
| BC060634.1 | - | 6 | 1212 | 6848779 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fibcd1| Gene Symbol: Fibcd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178887.4 | NP_849218.2 | 6 | 1212 | ||
| GenBank mRNA | AK046338.1 | - | 6 | 1174 | ||
| AK084379.1 | - | 7 | 1215 | |||
| BC060634.1 | - | 6 | 1212 | 6848779 | ||