siRNA Details
| Gene Symbol: Qser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123327.2 | NP_001116799.1 | 3 | 2771 | ||
| GenBank mRNA | AK078821.1 | - | 1 | 87 | ||
| AK141733.1 | - | 1 | 1082 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Qser1| Gene Symbol: Qser1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123327.2 | NP_001116799.1 | 3 | 2771 | ||
| GenBank mRNA | AK078821.1 | - | 1 | 87 | ||
| AK141733.1 | - | 1 | 1082 | |||